Cephradine ≥98%
Bottle
Catalog # APOSBUP17401-100MG
New Product
Supplier: Apollo Scientific
Cephradine ≥98%
Catalog # APOSBUP17401-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 38821-53-3
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
- CAS Number:38821-53-3
- Formula:C₁₆H₁₉N₃O₄S
- Molecular Weight:349,4 g/mol
Specifications
Specifications
- Pack type:Bottle



