Order Entry
Finland
Orders LinkContactUsLinkComponent
Ac4ManNAz ≥98%

Ac4ManNAz ≥98%

New Product
Supplier:   Apollo Scientific
Ac4ManNAz ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Ac4ManNAz ≥98%

3 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Ac4ManNAz
  • CAS Number:361154-30-5
  • Formula:C₁₆H₂₂N₄O₁₀
  • Molecular Weight:430,37 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 231,00 €
    231,00 €
    1 * 25 mg
       231,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 348,00 €
    348,00 €
    1 * 50 mg
       348,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 514,00 €
    514,00 €
    1 * 100 mg
       514,00 €

    Specifications:
    New Product
    New Product