Order Entry
Finland
Orders LinkContactUsLinkComponent
WEHL-04 ≥98%

WEHL-04 ≥98%

New Product
Supplier:   Apollo Scientific
WEHL-04 ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

4 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

  • CAS Number:114744-51-3
  • Formula:C₉H₁₀BrN
  • Molecular Weight:212,09 g/mol
Product Family Options

Product Information

  • Pakkaustyyppi
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 721,00 €
    721,00 €
    1 * 25 mg
       721,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 977,00 €
    977,00 €
    1 * 50 mg
       977,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 1 270,00 €
    1 270,00 €
    1 * 100 mg
       1 270,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 1 760,00 €
    1 760,00 €
    1 * 200 mg
       1 760,00 €

    Specifications:
    New Product
    New Product