Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Nonan-5-ol ≥98%

New Product
Supplier:   Apollo Scientific
Nonan-5-ol ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Nonan-5-ol ≥98%

3 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

  • CAS Number:623-93-8
  • Formula:C₉H₂₀O
  • Molecular Weight:144,25 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 400,00 €
    1 400,00 €
    1 * 25 mg
       1 400,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 930,00 €
    1 930,00 €
    1 * 50 mg
       1 930,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 2 550,00 €
    2 550,00 €
    1 * 100 mg
       2 550,00 €

    Specifications:
    New Product
    New Product