Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Lecithin ≥90%

New Product
Supplier:   Apollo Scientific
Lecithin ≥90%
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

  • CAS Number:8002-43-5
  • Formula:C₄₂H₈₀NO₈P
  • Molecular Weight:758,06 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 39,80 €
    39,80 €
    1 * 50 mg
       39,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 48,60 €
    48,60 €
    1 * 100 mg
       48,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 328,00 €
    328,00 €
    1 * 5 g
       328,00 €

    Specifications:
    New Product
    New Product