Order Entry
Finland
ContactUsLinkComponent
Cryovial for Long-Term Microorganism Preservation
Cryovial for Long-Term Microorganism Preservation
Catalog # LIOF27002
Supplier:  LIOFILCHEM
CAS Number:  
Cryovial for Long-Term Microorganism Preservation
Catalog # LIOF27002
Supplier:  LIOFILCHEM
CAS Number:  

Specifications

  • Pk:
    20
  • Description:
    Cryovial with 1,5 ml of tryptic soy broth + 15% glycerol, capacity: 50 vials

Specifications

About this item

Microbiology labs often require a straightforward method for long-term frozen storage of microorganisms for preserving key microbial strains without relying on frequent sub-culturing. Each vial contains tryptic soy broth +15% glycerol. This liquid guarantees the long-term frozen storage of microorganisms.

  • Cryopreservative medium- or long-term preservation of microorganisms and stock cultures

    Suitable for ultra-low temperature storage down to –70 °C

    Tested for sterility and growth performance

Long-term storage avoids risk of culture contamination and unwanted alterations in the organisms’ genetic and biochemical profiles.

Delivery:

Each package contains 20 tubes of 1,5 ml and detailed instruction sheet.