Ammonium Standard Solution Certified Reference Material
Catalog # 1.04622.0500
Supplier: Merck
New Product
CAS Number:
Specifications
- Pack type:Plastic bottle
- Description:Traceable to SRM from NIST NH₄Cl in H₂O 1000 mg/l NH₄ Certipur®
- Storage conditions:10 - 13 Other liquids and solids
- Storage temperature:Store at 15...+25 °C
- Pk Info:500 ml
Specifications
About this item
Density: 0,9993 g/cm3
WGK: NWG not water endangering
Certifications:
ISO 9001 & ACS