Specifications
- Capacity:60 L
- Colour:Clear
- Description:Clear, biohazard printed, with autoclave indicator
- Material:PP
- Thickness:50 µm
- Pk:200
- W×L:610×915 mm
Specifications
About this item
Sturdy, temperature resistant bags, ideal for disposing of non hazardous waste or hazardous waste that must first be autoclaved.
Autoclavable up to 135 °C maximum
200 mm twist ties included allows bag to expand and vent steam during cooling
Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709
-
Available with differnent thickness 37,5, 50 µm thick
Available with printed bag, which is printed with the universal biohazard symbol
Available with temperature indicator, text 'Contents Have Been Autoclaved' turns dark to indicates successful autoclaving
Highly resistant to tears and punctures
Can also be used for autoclaving other products. Supplied complete with wire closures that allow expansion of the bag and ventilation when cooling.
ISO 9001:2015, Sedex certified.