VWR® Autoclavable Bags, Polypropylene, Clear
Supplier: VWR Collection
Certificates
About this item
Sturdy, temperature resistant bags, ideal for disposing of non hazardous waste that must first be autoclaved.
Autoclavable up to 135 °C (275 °F) maximum
- 20 cm twist ties included allows bag to expand and vent steam during cooling
- Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709
- Available with differnent thickness 37,5 µm, 50 µm thick
- Available with plain or printed bag
- Highly resistant to tears and punctures
- Without autoclave Indicator
Can also be used for autoclaving other products. Supplied complete with wire closures that allow expansion of the bag and ventilation when cooling.
Certifications:
Certifications: ISO 9001:2015, Sedex.