Order Entry
Finland
ContactUsLinkComponent
SPINeasy® DNA Kit for Water
SPINeasy® DNA Kit for Water
Catalog # 116536050.
Supplier:  MP Biomedicals
New Product
CAS Number:  
SPINeasy® DNA Kit for Water
Catalog # 116536050.
Supplier:  MP Biomedicals
New Product
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Target molecule:
    DNA
  • Description:
    SPINeasy® DNA kit for water/gDNA extraction kit for water
  • Storage temperature:
    Store at room temperature (15...30 °C), dry environment
  • Contents:
    Components: 50×5 ml lysing matrix E, 60 ml lysis buffer W1, 8 ml lysis buffer W2, 550 μl RNase A solution, 15 ml inhibitor removal W, 30 ml binding buffer W, 9 ml wash buffer W1, 6 ml wash buffer W2, 10 ml DES buffer, 50X filter membrane, 50X column W1, 50×2,0 ml collection tubes, 50×1,5 ml Collection tubes
  • No. of reactions:
    50 preps

Specifications

About this item

SPINeasy DNA Kit for Water is a high-performance water genomic DNA (gDNA) extraction kit based on silica-membrane spin-column technology. This kit enables quick isolation of gDNA from water in less than 30 min. Water samples are processed using our uniquely formulated Lysis Buffer W1 and Lysing Matrix E to effectively lyse various types of cells. Column W1 provided in the kit has high binding capacity and selectivity for gDNA. The combination of components in the kit extracts gDNA of high yield and purity that is ready for downstream analyses such as PCR, restriction digestion and sequencing.

  • Quick Isolation of genomic DNA from water using silica membrane spin column technology

    For genomic research and environmental study

    Hazard Class 8

    Personal protective equipment: Safety goggles/face shield, gloves

    Sample type: Water samples including fresh and sea water, pond and river water, wastewater, etc.

Components: 50×5 ml lysing matrix E, 60 ml lysis buffer W1, 8 ml lysis buffer W2, 550 μl RNase A solution, 15 ml inhibitor removal W, 30 ml binding buffer W, 9 ml wash buffer W1, 6 ml wash buffer W2, 10 ml DES buffer, 50X filter membrane, 50X column W1, 50×2,0 ml collection tubes, 50×1,5 ml Collection tubes

Research Use Only (RUO).

Extract gDNA of high yield and purity from different types of water including river water, pond water, seawater, sewage that is ready for downstream analyses such as PCR, restriction digestion and sequencing.