Order Entry
Finland
ContactUsLinkComponent
Tork S2 Alcohol Hand Sanitiser Gel Mini
Tork S2 Alcohol Hand Sanitiser Gel Mini
  SCAA420102
 :  Essity
New Product
undefined
Tork S2 Alcohol Hand Sanitiser Gel Mini
  SCAA420102
 :  Essity
New Product
  • Description:
    Premium gel sanitiser, refill, 6× 475 ml
Tork alcohol hand sanitiser gel mini S2, perfume free with no sticky residue.
  • Effective against a variety of problem organisms, this hand sanitiser gel conforms with EN 1500, EN 12791 and EN 14476
  • Sanitise hands without leaving a sticky residue - this hand sanitiser gel is easy to spread
  • Improve hygiene in washrooms - the sealed refill with its single-use pump reduces the risk of cross-contamination
  • Reduce waste - the refill bottle collapses inside the hand sanitiser dispenser as the contents are used
  • Act against all enveloped viruses, rotavirus and norovirus
    Compatible with Tork S2 dispensers
Help prevent the spread of infections with our highly efficient Tork alcohol hand sanitiser gel. Effective against a variety of problem organisms, it even meets scrub room requirements, making it ideal for healthcare settings. Specially formulated for frequent use with no sticky build-up, this hand sanitiser gel contains moisturising ingredients. Use with the Tork S2 liquid soap dispenser, which is hygienic and easy to use.
 :
Use biocidal products safely. Always read the label and product information before use.