Order Entry
Finland
ContactUsLinkComponent
Occupational Shoes, Clogs, EASY 2.0 87621
Occupational Shoes, Clogs, EASY 2.0 87621
Catalog # ABEB87621/40
Supplier:  Abeba
New Product
Occupational Shoes, Clogs, EASY 2.0 87621
Catalog # ABEB87621/40
Supplier:  Abeba
New Product

Specifications

  • Shoe size:
    40
  • Colour:
    Black

Specifications

About this item

These shoes are ideal for professionals whose work requires constant movement and prolonged standing. The series low weight prevents leg fatigue. Shock Absorber technology cushions shocks and bumps, protecting against injuries and ensuring a high level of comfort - even on long workdays. Its outstanding performance parameters make the active series ideal for hospitals, nursing homes, and the food industry.

  • Increased breathability

    Easy to clean shaft

    Washable at 30°

    Resistance to acids and bases

    Resistance to short-term contact with hot substances up to 180 °C

    Upper material Microfiber

    Lining fabric

    Closure buckle

Lining - Increased water vapor permeability in the requirements of min 2 mg/(cm²×h) - Result is 14,6 mg/(cm²×h), the water vapor ratio is required at least 20 mg/cm² and the result is 117,2 mg/cm².

Shock absorber technology: Energy absorption in the heel area - according to the standard at least 20 J - result 29,4 J.

High abrasion resistance: Requirement of the standard: less than or equal to 150 mm³ - result is 46 mm³

Insole fabric, DGUV 112-191 orthopaedic insoles, replaceable insole. Outsole PU/PU, resistance to acids, resistance to bases, resistance to short-term contact with hot.

Substances up to 180 °C, PRT FLEX technology, shock absorber technology, self clean technology (self-cleaning outsole).

Certifications:

Standard EN ISO 20347:2012, OB, A, E, FO, SRC.