NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kits for the determination of ortho and total phosphate in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Coloured pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (OD) tubes. They are preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Delivery: Rugged box with 20 test tubes.
Specifications
- Model:MANA985081
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:455 g (including packaging)
Specifications
Related Information
Certificates
Search by Lot NumberIFU NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 1.pdf IFU NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 15.pdf IFU NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 45.pdf IFU NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 50.pdf IFU Ortho and Total Phosphate 5 Tube Test.pdf
More
Manual NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 1.pdf Manual NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 15.pdf Manual NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 45.pdf Manual NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 50.pdf Manual Ortho and Total Phosphate 5 Tube Test.pdf
Manual
More
NANOCOLOR® Photometric Tube Tests Ortho and Total Phosphate 15, 690 nm.pdf NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 1.pdf NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 15, 436 nm.pdf NANOCOLOR® Photometric Tube Tests Ortho- and Total-Phosphate 45.pdf Ortho and Total Phosphate 5 Tube Test.pdf
Additional Product Details
More