Order Entry
Finland
ContactUsLinkComponent
Vista Sterile Cleanroom Overboots
Vista Sterile Cleanroom Overboots
Catalog # ISOF80761
Supplier:  ISOFIELD IRELAND
New Product
Vista Sterile Cleanroom Overboots
Catalog # ISOF80761
Supplier:  ISOFIELD IRELAND
New Product
Specifications
  • Clothing Size:
    Universal
  • Sterile:
    +
  • Garment application:
    Cleanroom
  • Colour:
    White
  • Description:
    Vista cleanroom overboots
  • Cleanroom class:
    10/ISO 4; EU GMP (A)
Specifications
About this item
The Isofield Vista sterile cleanroom overboots are 50 cm (19,5”) tall and aseptically folded.
  • Sterile, cleanroom overboots
  • Extra-long design, 50 cm knee height
  • Two ties for added security
  • Anti-slip PU sole
  • Aseptically folded
Providing essential protection for GMP Grade A/B cleanroom applications, these anti-static, low-linting overboots are constructed with a non-slip PU sole, two sets of ties for secure fit, and excellent barrier properties.
Packaging:
Overboots are packaged 1 pair per inner bag, 10 pairs per outer bag, 8 outer bags per carton, 80 pairs per carton.