Order Entry
Finland
ContactUsLinkComponent
TMB ELISA Substrate (Slower Kinetic Rate)
TMB ELISA Substrate (Slower Kinetic Rate)
Supplier:  Abcam
Certificates
undefined
TMB ELISA Substrate (Slower Kinetic Rate)
Supplier:  Abcam

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Storage temperature:
    Shipped at 4 °C. Store at 4 °C short term (1 - 2 weeks). Store at 4 °C. Please see notes section.
  • Regulatory status:
    RUO

Specifications

Frequently Bought Together