Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Norepinephrine ELISA Kit

Norepinephrine ELISA Kit, 96 tests, 96 Tests
  ABCAAB287789-96
 :   Abcam
Norepinephrine ELISA Kit
  ABCAAB287789-96
 :  Abcam
 :  
Norepinephrine ELISA Kit
Norepinephrine ELISA Kit

Norepinephrine ELISA Kit, 96 tests, 96 Tests

 :  
 
872,00 €
   
1 * 1 Each
 

Norepinephrine ELISA kit is a competitive ELISA designed to quantify norepinephrine/noradrenaline (NA) with a sensitivity of <9.375 pg/ml. - Wide dynamic range - quantifies 15.625 - 1000 pg/ml. Broad sample compatibility - works with serum, plasma, tissue homogenates, cell culture supernatants, and other biological fluids. Assay platform - microplate (12×8-well strips) for easy setup and reliable results.

 

 

  • Pk:
    96 Tests
  • Description:
    Norepinephrine ELISA Kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB287789-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    9,375 pg/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Sample type:
    Plasma, Tissue Homogenate, Cell culture supernatant, Serum, Other biological fluids
  • Target protein:
    Noradrenaline
  • Detection range:
    15,625 - 1000 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Assay Type:
    Competitive