Order Entry
Finland
ContactUsLinkComponent
Mouse CXCL11 ELISA Kit (I-TAC)
Mouse CXCL11 ELISA Kit (I-TAC)
Catalog # ABCAAB204519-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse CXCL11 ELISA Kit (I-TAC)
Catalog # ABCAAB204519-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Mouse CXCL11 ELISA kit (I-TAC)
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse CXCL11 ELISA kit (I-TAC) is a single-wash 90-min Simplestep used to quantify Mouse CXCL11 (I-TAC) with a sensitivity of 1,4 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.