Anti-IgG Rabbit Monoclonal Antibody [clone: EPR25A] (Alexa Fluor® 488)
Catalog # ABCAAB199091-100
Supplier: Abcam
Specifications
- Size:100 µl
- Pack type:Vial
Specifications
About this item
Alexa Fluor® 488 Rabbit IgG, monoclonal [EPR25A] - Isotype Control.
Type: Control
Antigen: IgG
Clonality: Monoclonal
Clone: EPR25A
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: