Order Entry
Finland
ContactUsLinkComponent
Digital thermo-hygrometer, TFH 620
Digital thermo-hygrometer, TFH 620
  620-1394
 :  EBRO
undefined
Digital thermo-hygrometer, TFH 620
  620-1394
 :  EBRO
  • Description:
    TFH 620 with air probe TPH 100
  • Accuracy:
    ±0,3 °C; ±2,0 % HR
  • Pk:
    1
This robust and impact resistant thermo-hygrometer provides high accuracy measurements for monitoring storage checks.
  • Automatic deactivation
  • Capacitive humidity sensor
  • Two measurement channels
Thermistor sensor.
 : Supplied with an internal sensor, a six-point factory calibration certificate, a lithium three-volt battery for up to five years of life with a charge indicator.