Capillary columns, PERMABOND® CW 20 M
Supplier: MACHEREY-NAGEL
Certificates
About this item
Polar GC phase.
- Suitable for solvent analysis
- Maximum temperature for isothermal operation 220 °C (0,53 mm Int.Ø columns: 200 °C)
- Maximum temperature for short isotherms in a temperature program 240 °C (0,53 mm Int.Ø columns: 220 °C)
Polyethylene glycol (20 kD) phase.
USP code: G16.