Capillary columns, OPTIMA® 5 Amine
Special GC phase for the analysis of amines.
- Especially deactivated for the analysis of polyfunctional amines such as ethanolamines, amino-functionalized diols and similar compounds
- Improved linearity for analyses of active components at trace levels
- Max. temperature for isothermal operation 300 °C
- Max. temperature for short isotherms in a temperature program 320 °C
USP code: G27 / G36.