Capillary columns, OPTIMA® 240
Medium polar GC phase.
- Recommended for FAMEs, and dioxins
- Maximum temperature for isothermal operation 260 °C
- Maximum temperature for short isotherms in a temperature program 280 °C
33% cyanopropyl-methyl – 67% dimethylpolysiloxane phase.