
Isoscopoletin (6-hydroxy-4-methylcoumarin) ≥98.0% (by GC, titration analysis)
Catalog # TCIAH1005-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:246.0 to 250.0 deg-C
Specification Test Results
About this item
:
:176.17 g/mol
EINECS:219-146-3
:GHS07
CAS Number:2373-31-1
Formula:C₁₀H₈O₃
MDL Number:MFCD00016967