
Methyl-3-(bromomethyl)benzoate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1129-28-8
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00051437
Boiling Pt:169°C
EINECS:419-100-4
UN:3261
ADR:8,III
:229.07 g/mol
:GHS05
Specification Test Results
- Appearance:White or Colorles to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Melting point:43.0 to 47.0 deg-C
- Solubility in Methanol:almost transparency