Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-Jasmone
cis-Jasmone ≥92.0% (by GC)
cis-Jasmone ≥92.0% (by GC)
:
TCI
cis-Jasmone ≥92.0% (by GC)
:
TCI
:
488-10-8
:
:
C₁₁H₁₆O
:
MFCD00001402
:
14,05259
:
128°C
:
164.25 g/mol
:
207-668-4
:
GHS07
Appearance:
Colorless to Light orange to Yellow clear liquid
Purity (GC):
min. 92.0 %
Manual Bioscience and Analyticalscience