
Dibismuth trisulphate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7787-68-0
:
Formula:Bi₂O₁₂S₃
MDL Number:MFCD00036140
:706.13 g/mol
Merck Index:14,1286
Melting Pt:405 °C (dec.)
Density:5.08
EINECS:232-129-5
:GHS07
Storage Temperature:Ambient