Order Entry
Finland
ContactUsLinkComponent
Nicotinoyl chloride hydrochloride for synthesis
Nicotinoyl chloride hydrochloride for synthesis
Catalog # 8.21552.0250
Supplier:  Merck
CAS Number:  
Nicotinoyl chloride hydrochloride for synthesis
Catalog # 8.21552.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :178.02 g/mol
EINECS:243-648-1
UN:3261
ADR:8,II
CAS Number:20260-53-1
Formula:C₆H₅Cl₂NO
MDL Number:MFCD00012820