Order Entry
Finland
ContactUsLinkComponent
Luminol (3-Aminophthalhydrazide) ≥98.0% (by HPLC, titration analysis) chemiluminescence reagent
Luminol (3-Aminophthalhydrazide) ≥98.0% (by HPLC, titration analysis) chemiluminescence reagent
Supplier:  TCI
Certificates
undefined
Luminol (3-Aminophthalhydrazide) ≥98.0% (by HPLC, titration analysis) chemiluminescence reagent
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Solubility in Na2CO3 (100g/L):
    almost transparency
  • Drying loss:
    max. 0.5 %

Specification Test Results

Related Information