Order Entry
Finland
ContactUsLinkComponent
1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate ≥96.0% (by total nitrogen basis)
Corrosive
Irritant
1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate ≥96.0% (by total nitrogen basis)
Catalog # TCIAD3621-1G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate ≥96.0% (by total nitrogen basis)
Catalog # TCIAD3621-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 96.0 %

Specification Test Results

About this item

 :
 :424.33 g/mol
UN:1759
ADR:8,III
 :GHS05
 :GHS07
CAS Number:286014-42-4
Formula:C₂₃H₃₃BF₄N₂
MDL Number:MFCD04973311