

2,5-Dihydro-2,5-dimethoxyfuran (mixture of cis and trans isomers) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAD1759-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:130.14 g/mol
Flash Pt:47°C
EINECS:206-367-5
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:332-77-4
Formula:C₆H₁₀O₃
MDL Number:MFCD00003220