Octaethylene glycol monododecylether (C12E8) ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3055-98-9
:
Formula:C₂₈H₅₈O₉
MDL Number:MFCD00043234
:538.754 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 97.0 %