3-(Trimethylsilyl)-2-oxazolidinone ≥98.0% (by GC) trimethylsilylating reagent
TCIAT1535-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Purity (GC):min. 98.0 %
:
:159.26 g/mol
:122°C
:76°C
:Freezer
:256-098-2
:43112-38-5
:C₆H₁₃NO₂Si
:MFCD00010197