


Tetrahydrofuran ≥99.5% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Merck Index:14,09211
Boiling Pt:65°C
Melting Pt:–108°C
Density:0,888 g/cm³ (20 °C)
Flash Pt:–15°C
UN:2056
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Storage Temperature:Ambient
EINECS:203-726-8
REACH:05-2114615188-48
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.5 %
- Water:max. 0.05 %
- Peroxide:max. 0.005% (as H2O2)
Specification Test Results
Frequently Bought Together