
4-Methoxybenzyl chloride ≥98.0% (by GC, titration analysis) stabilised
: TCI
:824-94-2
:
:C₈H₉ClO
:MFCD00000915
:156.61 g/mol
:127°C
:–1 °C
:1,072 g/cm³ (0 °C)
:109°C
:3265
:8,II
:Stabilised with amylene
:212-540-6
:GHS05
:Refrigerator
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Frequently Bought Together