
4-Amino-3-nitrophenylboronic acid pinacol ester ≥98.0% (by GC)
Catalog # TCIAN0824-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:175.0 to 180.0 deg-C
Specification Test Results
About this item
:
:264.09 g/mol
:GHS07
CAS Number:833486-94-5
Formula:C₁₂H₁₇BN₂O₄
MDL Number:MFCD06795680