

2-(4-Chlorophenoxy)ethanol ≥98.0% (by GC)
Catalog # TCIAC0251-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White or Colorless to Yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Freezing point:28.0 to 31.0 deg-C
Specification Test Results
About this item
:
:172.61 g/mol
Boiling Pt:187°C
Melting Pt:31…32 °C
Density:1,2515 g/cm³ (20 °C)
EINECS:217-578-7
UN:2811
ADR:6.1,III
:GHS05
:GHS06
CAS Number:1892-43-9
Formula:C₈H₉ClO₂
MDL Number:MFCD00014036