

Allylchlorodimethylsilane ≥96.0% (by GC) allyldimethylsilylating reagent for GC/MS
Supplier: TCI
Certificates
About this item
CAS Number:4028-23-3
:
Formula:C₅H₁₁ClSi
MDL Number:MFCD00000506
:134.68 g/mol
Boiling Pt:113°C
Flash Pt:5°C
Storage Temperature:Refrigerator
EINECS:223-706-2
UN:2985
ADR:3,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 96.0 %