10% SDS (Sodium Dodecyl Sulfate), Corning®
Catalog # MDTC46-040-CI
Supplier: Corning
CAS Number:
Specifications
- Pk:1
- Description:10% SDS (Sodium Dodecyl Sulfate)
- Storage temperature:Ambient
- Shipping temperature:Ambient
- Pk Info:100 ml
Specifications
About this item
Sodium Dodecyl Sulfate (SDS) is an anionic detergent with applications in SDS-PAGE protein analysis and nucleic acid purification. It is a wetting agent that is effective in both acid and alkaline solutions, and is most often used as a protein and lipid solubilization reagent. SDS is also a powerful protein denaturant. This 10% solution is prepared in 18 megaohm water and is 0.2µm filtered.