Coomassie® brilliant blue R-250 for electrophoresis
Catalog # TCIAB3194-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark purple powder to crystal
- Lambda max.:586.0 to 590.0 nm (MeOH)
- Absorbance (E1%1cm):min. 800 (MeOH, 586.0 to 590.0 nm)
- Functionality test:to pass test
Specification Test Results
About this item
:
:825.98 g/mol
Melting Pt:174…180 °C
Storage Temperature:Ambient
EINECS:228-060-5
CAS Number:6104-59-2
Formula:C₄₅H₄₄N₃NaO₇S₂
MDL Number:MFCD00041762