
2-Chloro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:182344-18-9
:
Formula:C₇H₅BClF₃O₂
MDL Number:MFCD00797335
:224.37
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 109.0 %