2-(4-Bromophenyl)ethanol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4654-39-1
:
Formula:C₈H₉BrO
MDL Number:MFCD00002897
:201.06 g/mol
Boiling Pt:150°C
Melting Pt:36…38 °C
Density:1,4678 g/cm³ (28.5 °C)
EINECS:225-093-7
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together