
2,4,5-Trifluoro-4-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:116751-24-7
:
Formula:C₇H₃F₃O₃
MDL Number:MFCD00800378
:192.09 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:144.0 to 148.0 deg-C