3-Methoxy-3-methyl-1-butanol ≥98.0% (by GC)
: TCI
:56539-66-3
:
:C₆H₁₄O₂
:MFCD00044771
:118.18 g/mol
:174°C
:–50 °C
:0,9038 g/cm³ (20 °C)
:67°C
:Ambient
:260-252-4
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together