Order Entry
Finland
ContactUsLinkComponent
3-Mercapto-1,2-propanediol ≥98.0% (by GC, titration analysis) matrix for FABMS and liquid SIMS
Toxic
3-Mercapto-1,2-propanediol ≥98.0% (by GC, titration analysis) matrix for FABMS and liquid SIMS
Supplier:  TCI
Certificates
undefined
Toxic
3-Mercapto-1,2-propanediol ≥98.0% (by GC, titration analysis) matrix for FABMS and liquid SIMS
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Iodometric back titration):
    min. 98.0 %

Specification Test Results

Related Information