Order Entry
Finland
ContactUsLinkComponent
1,4-Cyclohexanediol cis- and trans-mixture ≥99.0% (by GC)
1,4-Cyclohexanediol cis- and trans-mixture ≥99.0% (by GC)
Supplier:  TCI
Certificates
undefined
1,4-Cyclohexanediol cis- and trans-mixture ≥99.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 % (total of isomers)
  • Melting point:
    100.0 to 104.0 deg-C
  • Solubility in hot Acetone:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together