

Dimethyl sulphide ≥99.0% (by GC)
: TCI
:75-18-3
:
:C₂H₆S
:MFCD00008562
:62.14 g/mol
:14,06123
:37°C
:–98 °C
:0,85 g/cm³ (20 °C)
:–34°C
:Refrigerator
:200-846-2
:1164
:3,II
:GHS02
:GHS05
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %