Glycine ≥99.0% (by titrimetric analysis) for electrophoresis
: TCI
:56-40-6
:
:NH₂CH₂COOH
:MFCD00008131
:75.07 g/mol
:14,04491
:181 °C (1013 hPa)
:240°C
:1,64 g/cm³ (20 °C)
:Ambient
:200-272-2
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Functionality test:to pass test