




Dekahydronaftaleeni (cis- ja trans-isomeerien seos) cis- and trans-mixture ≥99.0% (by GC)
Catalog # TCIAD1738-500ML
Supplier: TCI
CAS Number:
Specifications
- Pak.:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0% (total of isomer)
- Content of sulfur:max. 1.0 ppm
Specification Test Results
About this item
:
:138.25 g/mol
Merck Index:14,02846
Boiling Pt:190°C
Melting Pt:∼–40 °C
Density:0,8696 g/cm³ (20 °C)
Flash Pt:58°C
Storage Temperature:Huoneenlämpö
EINECS:202-046-9
UN:1147
ADR:3,III
:GHS02
:GHS05
:GHS06
:GHS08
:GHS09
CAS Number:91-17-8
Formula:C₁₀H₁₈
MDL Number:MFCD00004130