Order Entry
Finland
ContactUsLinkComponent
Guanidinium thiocyanate ≥98.0% (by titrimetric analysis) for biochemical research
Guanidinium thiocyanate ≥98.0% (by titrimetric analysis) for biochemical research
Supplier:  TCI
Certificates
undefined
Guanidinium thiocyanate ≥98.0% (by titrimetric analysis) for biochemical research
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    118.0 to 122.0 deg-C
  • Suitability for protein-analysis:
    to pass test
  • Protein denaturation test:
    to pass test

Specification Test Results

Related Information