
2-(Trifluoromethoxy)phenylboronic acid (contains varying amounts of Anhydride)
: TCI
:175676-65-0
:
:C₇H₆BF₃O₃
:MFCD01320763
:205.93 g/mol
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):97.0 to 110.0 %
Frequently Bought Together